Skip to Content
Merck
All Photos(1)

Documents

SAE0003

Sigma-Aldrich

4-Hydroxyphenylpyruvate Dioxygenase from Streptomyces avermitilis

recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

liquid

specific activity

≥5 U/mg

mol wt

~41.8 kDa

shipped in

dry ice

storage temp.

−70°C

General description

4-Hydroxyphenylpyruvate Dioxygenase (4-HPPD) is a key enzyme in the catabolism of tyrosine and phenylalanine. 4-HPPD catalyzes the synthesis of homogenistate (2,5-dihydroxyphenylacetate) from ρ-hydroxyphenylpyruvate and molecular oxygen. 4-HPPD also catalyzes the conversion of α-ketoisocaproate to Β-hydroxyisovalerate. 4-HPPD requires Fe2+ as an essential cofactor.

Rat 4-HPPD has a minimum of 80% homology with human 4-HPPD. Rat 4-HPPD is expressed abundantly in hepatocytes. It is also expressed in kidney, particularly in epithelial cells of the proximal tubules and the distant tubules. 4-HPPD has been identified as a potential biomarker of drug-induced liver injury in rat studies.

Unit Definition

One unit will oxidize 1 μmole of molecular oxygen per minute at pH 7.0, 25 °C, in the presence of 4-hydroxyphenylpyruvate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service