Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702229

Sigma-Aldrich

Anti-PSD93/chapsyn-110/DLG2 Antibody, clone 0G4O6, Rabbit Monoclonal

Synonym(s):

PPP1R58, PSD-93, PSD93, chapsyn-110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

0G4O6, monoclonal

form

liquid

mol wt

110 kDa

species reactivity

rat, human, rat

concentration

1.20 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

RRVERKERARLKTVKFNAKPGVIDSKGSFNDKRKKSFIFSRKFPFYKNKEQSEQETSDPERGQEDLILSYEPVTRQEINYTRPVIILGPMKDRINDDLISE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLG2(1740)

General description

This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. The encoded protein forms a heterodimer with a related family member that may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. Multiple transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been described, but their full-length nature is not known. [provided by RefSeq, Dec 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 600-700 of human PSD93/chapsyn-110/DLG2 (Q15700).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service