Skip to Content
Merck
All Photos(1)

Key Documents

SAB5701796

Sigma-Aldrich

Anti-Phospho-ULK1-S555 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,896.00

R$2,896.00


Estimated to ship onMay 01, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB2227


Select a Size

Change View
100 μL
R$2,896.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,896.00


Estimated to ship onMay 01, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB2227

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human, rat, mouse

concentration

2.81 mg/mL

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer555)

Gene Information

human ... ULk1(8408)

General description

Serine/threonine-protein kinase involved in autophagy in response to starvation. Acts upstream of phosphatidylinositol 3-kinase PIK3C3 to regulate the formation of autophagophores, the precursors of autophagosomes. Part of regulatory feedback loops in autophagy: acts both as a downstream effector and negative regulator of mammalian target of rapamycin complex 1 (mTORC1) via interaction with RPTOR. Activated via phosphorylation by AMPK and also acts as a regulator of AMPK by mediating phosphorylation of AMPK subunits PRKAA1, PRKAB2 and PRKAG1, leading to negatively regulate AMPK activity. May phosphorylate ATG13/KIAA0652 and RPTOR; however such data need additional evidences. Plays a role early in neuronal differentiation and is required for granule cell axon formation. May also phosphorylate SESN2 and SQSTM1 to regulate autophagy (PubMed:25040165).

Immunogen

A synthetic phosphorylated peptide around S555 of human Phospho-ULK1-S555 (NP_003556.1).

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service