Skip to Content
Merck
All Photos(2)

Key Documents

SAB5701118

Sigma-Aldrich

Anti-CEACAM5 antibody produced in mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,669.00

R$3,669.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,669.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,669.00


Check Cart for Availability

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

mouse, human

concentration

2.17 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:5000-1:10000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEACAM5(1048)

General description

This gene encodes a cell surface glycoprotein that represents the founding member of the carcinoembryonic antigen (CEA) family of proteins. The encoded protein is used as a clinical biomarker for gastrointestinal cancers and may promote tumor development through its role as a cell adhesion molecule. Additionally, the encoded protein may regulate differentiation, apoptosis, and cell polarity. This gene is present in a CEA family gene cluster on chromosome 19. Alternative splicing results in multiple transcript variants.

Immunogen

Recombinant protein of human CEACAM5

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I wonder which amino acid sequence was used as an antigen. Thanks,

    1 answer
    1. The immunogen used to make this monoclonal antibody is the mature CEACAM5 protein. This does not include the N-terminal signal peptide, nor the C-terminal propeptide. The epitope to which the antibody binds has not been mapped.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service