Skip to Content
Merck
All Photos(1)

Key Documents

SAB5700386

Sigma-Aldrich

Anti-Phospho-AURKA-T288 antibody produced in rabbit

Synonym(s):

AIK, ARK1, AURA, AURKA, BTAK, PPP1R47, STK15, STK6, STK7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,011.00

R$3,011.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,011.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,011.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45

species reactivity

human

concentration

3.55 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr288)

Gene Information

human ... AURKA(6790)

General description

The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene.

Immunogen

A synthetic phosphorylated peptide around T288 of human AURKA (NP_003591.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service