Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600071

Sigma-Aldrich

Anti-alpha-Cardiac Actin (ACTC1) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM257, purified immunoglobulin

Synonym(s):

Actin, alpha cardiac muscle 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,966.00

R$2,966.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,966.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,966.00


Check Cart for Availability

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM257, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:1000-1:2000
immunohistochemistry: 1:1000-1:2000

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTC1(70)

Specificity

The antibody reacts to human and mouse alpha cardiac actin (Actin, alpha cardiac muscle 1). This antibody may also react to bovine or rat alpha-cardiac actin, as predicted by immunogen homology

Immunogen

Synthetic peptide corresponding to the N-terminus of human alpha-cardiac actin

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kittiya Islam et al.
Cell adhesion & migration, 15(1), 152-165 (2021-05-21)
Extensive desmoplasia in cholangiocarcinoma (CCA) is associated with tumor aggressiveness, indicating a need for further understanding of CCA cell-matrix interaction. This study demonstrated laminin as the most potent attractant for CCA cell migration and the vast elevation of its receptor
Lynn Yap et al.
NPJ Regenerative medicine, 8(1), 26-26 (2023-05-27)
Ischemic heart disease, which is often associated with irreversibly damaged heart muscle, is a major global health burden. Here, we report the potential of stem cell-derived committed cardiac progenitors (CCPs) have in regenerative cardiology. Human pluripotent embryonic stem cells were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service