Skip to Content
Merck
All Photos(2)

Key Documents

SAB5202454

Sigma-Aldrich

Monoclonal Anti-4-Hydroxy-2-hexenal antibody produced in mouse

clone 6F10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$1,470.00

R$1,470.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$1,470.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,470.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F10, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1.0 mg/mL

technique(s)

flow cytometry: 1:50
immunofluorescence: 1:50
western blot: 1:1000

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

hydroxylation (4-Hydroxy-2-hexenal)

Related Categories

Immunogen

Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service