SAB5201832
Monoclonal Anti-ENaC alpha-Atto 700 antibody produced in mouse
clone 14E10
About This Item
biological source
mouse
conjugate
Atto 700 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
14E10, monoclonal
form
buffered aqueous solution
mol wt
85kDa
species reactivity
mouse, rat
concentration
1.0 mg/mL
technique(s)
immunohistochemistry: 1:100
western blot: 1:1000
isotype
IgG1
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Scnn1a(25122)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service