Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702056

Monoclonal Anti-CD63 PerCP produced in mouse

new

Synonym(s):

Granulophysin, LAMP-3, ME491, MLA1, OMA81H, TSPAN30, Tetraspanin-30, Tspan-30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

Peridinin-Chlorophyll-Protein Complex conjugate

Quality Level

antibody product type

primary antibodies

clone

MEM-259, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD63(967)

General description

CD63 (LAMP-3, lysosome-associated membrane protein-3), a glycoprotein of tetraspanin family, is present in late endosomes, lysosomes and secretory vesicles of various cell types. It is also present in the plasma membrane, usually following cell activation. Hence, it has become an widely used basophil activation marker. In mast cells, however, CD63 exposition does not need their activation. CD63 interacts with integrins and affects phagocytosis and cell migration, it is also involved in H/K-ATPase trafficking regulation of ROMK1 channels. CD63 also serves as a T-cell costimulation molecule. Expression of CD63 can be used for predicting the prognosis in earlier stages of carcinomas.

Specificity

The antibody MEM-259 reacts with an extracellular/luminal epitope of CD63 (LAMP-3), a 40-60 kDa tetraspan glycoprotein expressed by granulocytes, platelets, T cells, monocytes/macrophages and endothelial cells. Cell surface exposition of CD63 is usually activation-dependent.

Immunogen

HPB-ALL T cell line

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated Peridinin-Chlorophyll Protein (PerCP) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service