Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700934

Sigma-Aldrich

Monoclonal Anti-CD222/IGF2 receptor/CIMPR antibody produced in mouse

clone MEM-238

Synonym(s):

CIMPR, IGF2R, M6P-R, MPR1, MPR300

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,612.00

R$2,612.00


Estimated to ship onMay 06, 2025



Select a Size

Change View
100 μG
R$2,612.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$2,612.00


Estimated to ship onMay 06, 2025


biological source

mouse

antibody product type

primary antibodies

clone

MEM-238, monoclonal

form

buffered aqueous solution

species reactivity

nonhuman primates, human

concentration

1 mg/mL

technique(s)

flow cytometry: 4 μg/mL

accession no.

P11717

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGF2R(3482)

General description

The antibody MEM-238 recognizes an epitope between amino acids 192-697 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal compartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.

Immunogen

Recombinant Vaccinia virus encoding CD222.

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service