Skip to Content
Merck
All Photos(1)

Documents

SAB4700759

Sigma-Aldrich

IgG3 Isotype Control murine

clone PPV-07, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.42

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

PPV-07, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG3

shipped in

wet ice

storage temp.

2-8°C

General description

This mouse IgG3 monoclonal antibody (clone PPV-07) reacts with undefined epitope on a plant pathogen.
The specificity of staining by monoclonal antibodies to target antigens should be verified by establishing the amount of non-specific antibody binding. Especially at higher concentration (more than 15 μg/ml) the antibody staining usually has consignable background. To this end a non-reactive immunoglobulin of the same isotype is included as a negative control for each specific monoclonal antibody used in a particular immunoassay. The monoclonal antibody PPV-07, generated against a plant pathogen, does not cross-react with other species, and hence all the background that could be observed when working with this antibody would be a result of general nonspecific interactions between an mouse IgG3 molecule and the respective sample under the particular conditions.

Immunogen

Plum pox virus

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Maresch et al.
Proteomics, 16(9), 1321-1330 (2016-03-10)
With mice being the top model organism in immunology and with Fc glycosylation being increasingly recognized as important modulator of antibody function, the time has come to take a look at the glycosylation of mouse IgG isotypes. Tryptic glycopeptides of
George C Brittain et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 91(4), 351-363 (2017-04-04)
A biochemical system and method have been developed to enable the quantitative measurement of cytoplasmic versus nuclear localization within cells in whole blood. Compared with the analyses of nuclear localization by western blot or fluorescence microscopy, this system saves a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service