Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700562

Sigma-Aldrich

Monoclonal Anti-Cd86-FITC antibody produced in rat

clone GL-1, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rat

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GL-1, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

mouse ... Cd86(12524)

General description

Cluster of differentiation 86 (CD86), also known as B7-2, is encoded by the gene mapped to human chromosome 3q13.33. CD86 is a co-stimulatory molecule expressed on various immune cells. The encoded protein belongs to the immunoglobulin (Ig) superfamily.
The rat monoclonal antibody GL-1 reacts with CD86 (B7-2), a 70-80 kDa type I transmembrane glycoprotein of immunoglobulin supergene family, expressed on professional antigen-presenting cells, such as dendritic cells, macrophages or activated B lymphocytes.

Immunogen

LPS-activated CBA/Cs mouse splenic B cells

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Cluster of differentiation 86 (CD86) plays a vital role in immune responses. Mutations in the gene leads to the development of pneumonia-induced sepsis by downregulating CD86 gene expression in monocytes. Additionally, genetic variation in the gene increases the risk of susceptibility to colorectal cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MicroRNA binding site polymorphisms as biomarkers of cancer risk.
Pelletier C and Weidhaas JB
Expert Review of Molecular Diagnostics, 10, 817-829 (2010)
Distinct role of CD86 polymorphisms (rs1129055, rs17281995) in risk of cancer: evidence from a meta-analysis.
Geng P
PLoS ONE, 9 (2014)
CD86 polymorphism affects pneumonia-induced sepsis by decreasing gene expression in monocytes.
Song H
Inflammation, 38, 879-885 (2015)
B7-H3: a costimulatory molecule for T cell activation and IFN-gamma production.
Chapoval Al
Nature Immunology, 2, 269-274 (2001)
Wen-Horng Wang et al.
ImmunoHorizons, 3(7), 254-261 (2019-07-30)
The SYK protein-tyrosine kinase is a well-known mediator of signals elicited by the clustering of BCR complexes and other receptors that bear components that contain one or more ITAM sequences. Additional roles for the kinase in signaling through other receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service