Skip to Content
Merck
All Photos(1)

Documents

SAB4700188

Sigma-Aldrich

Monoclonal Anti-CD44 antibody produced in rat

clone IM7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Pgp-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IM7, monoclonal

form

buffered aqueous solution

species reactivity

mouse, horse, canine, human, feline

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

General description

The CD44 (cell-surface glycoprotein) gene with 20 exons is mapped to human chromosome 11p13. The gene codes for a transmembrane glycoprotein.
The rat monoclonal antibody IM7 reacts with CD44 antigen (Phagocyte glycoprotein 1), an 80-95 kDa transmembrane glycoprotein (hyaladherin family) present on the most of cells and tissues (leukocytes, endothelial cells, mesenchymal cells, etc.); it is negative on platelets and hepatocytes. The antibody reacts with all isoforms of mouse CD44.

Immunogen

Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1

Application

Monoclonal Anti-CD44 antibody produced in rat has been used in immunohistochemistry.
The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

CD44 (cell-surface glycoprotein) is involved in cell-cell and cell-extracellular matrix interactions. It plays an important role in lymphocyte homing and lymphocyte activation. It also acts as a metastasis suppressor gene for prostatic cancer. Overexpression of CD44 leads to esophageal squamous cell carcinoma (ESCC). CD44 inhibits macrophage multinucleation by interacting with its ligands, hyaluronic acid, chondroitin sulfates and osteopontin. 100kDa form of CD44, facilitates the binding of poliovirus to HeLa cells. In addition, it also implicated in the infection of mononuclear phagocytes by human immunodeficiency virus (HIV).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rare missense variant in RCL1 segregates with depression in extended families
Amin N
Molecular Psychiatry (2017)
Guoliang Zhang et al.
Acta biochimica et biophysica Sinica, 46(7), 540-547 (2014-05-23)
Differentiation of monocytes into macrophages is an important process under physiological and pathological conditions, but the underlying mechanism of monocyte differentiation is not completely clear. Some adhesion molecules have been reported to play an important role in cell differentiation. CD44
Heli Li et al.
Journal of immunology research, 2018, 4807145-4807145 (2019-01-24)
The immunologic interaction between parenchyma cells and encircling inflammatory cells is thought to be the most important mechanism of biliary damage and repair in primary sclerosing cholangitis (PSC). Monocytes/macrophages as master regulators of hepatic inflammation have been demonstrated to contribute
Gongcai Qiu et al.
International journal of oncology, 55(1), 45-58 (2019-05-07)
The present study aimed to explore the long non‑coding RNA (lncRNA) expression profiles and correlation of lnc‑PKD2‑2‑3 with tumor features and prognosis, and to investigate its effect on regulating cancer‑cell stemness and its potential as a cancer stem cell (CSC) marker
CD44 occupancy prevents macrophage multinucleation.
Sterling H
The Journal of Cell Biology, 143, 837-847 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service