Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700014

Sigma-Aldrich

Monoclonal Anti-B2M antibody produced in mouse

clone B2M-02, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-β-2-microglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,498.00

R$2,498.00


In StockDetails



Select a Size

Change View
100 μG
R$2,498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,498.00


In StockDetails


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B2M-02, monoclonal

form

buffered aqueous solution

species reactivity

human, pig

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... B2M(567)

General description

β2-microglobulin (B2M) is a non-glycosylated polypeptide composed of 99 amino acids. It is present on the surface of all nucleated cells. The protein is characterized with the single polypeptide chain, which is linked noncovalently to major histocompatibility complex (MHC) class I cell surface antigen. The gene coding for B2M is mapped to human chromosome 15q.

Immunogen

Purified isolated human beta2-microglobulin

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

β2-microglobulin (B2M) plays a vital role in cell survival, proliferation and metastasis in various types of cancer. Elevated concentration of β-2-microglobulin (B2M) can be considered as a prognostic marker for hemophagocytic lymphohistiocytosis (HLH). Elevated concentration of circulating B2M, due to renal dysfunction and long term hemodialysis leads to dialysis-related amyloidosis. Overexpression of serum B2M is observed in hepatitis C virus (HCV) related chronic liver diseases, therefore it might act as a potential marker for the HCV disease progression towards cirrhosis and carcinoma. Missense mutation in the B2M gene results in familial hypercatabolic hypoproteinemia (FHH).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Nadeau et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(3), 814-818 (1984-02-01)
Linkage relationships of homologous loci in man and mouse were used to estimate the mean length of autosomal segments conserved during evolution. Comparison of the locations of greater than 83 homologous loci revealed 13 conserved segments. Map distances between the
S M Ouda et al.
Asian Pacific journal of cancer prevention : APJCP, 16(17), 7825-7829 (2015-12-03)
Egypt has the highest prevalence of HCV infection in the world (~14.7%). Around 10-15% of HCV-infected persons will advance to cirrhosis within the first 20 years. The incidence of HCC is expected to grow in the next two decades, largely
Zoe Hall et al.
The Journal of biological chemistry, 291(9), 4626-4637 (2015-12-15)
β2-Microglobulin (β2m), a key component of the major histocompatibility class I complex, can aggregate into fibrils with severe clinical consequences. As such, investigating the structural aspects of the formation of oligomeric intermediates of β2m and their subsequent progression toward fibrillar
Tiantong Jiang et al.
Disease markers, 2016, 1523959-1523959 (2016-04-26)
To determine the prognostic significance of beta2 microglobulin (β2-m) concentrations in patients with hemophagocytic lymphohistiocytosis (HLH), a rare disorder caused by pathologic activation of the immune system. The study population consisted of 74 patients diagnosed with HLH and 35 healthy
Jonghan Kim et al.
Clinical immunology (Orlando, Fla.), 122(2), 146-155 (2006-10-19)
The nonclassical MHC class-I molecule, FcRn, salvages both IgG and albumin from degradation. Here we introduce a mechanism-based kinetic model for human to quantify FcRn-mediated recycling of both ligands based on saturable kinetics and data from the literature using easily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service