Skip to Content
Merck
All Photos(1)

Key Documents

SAB4600048

Sigma-Aldrich

Monoclonal Anti-6X His IgG, CF 488A antibody produced in mouse

~2 mg/mL, clone 33D10.D2.G8, purified immunoglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
R$2,611.00

R$2,611.00


Check Cart for Availability


Select a Size

Change View
50 μL
R$2,611.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

R$2,611.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

CF 488A conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

33D10.D2.G8, monoclonal

form

buffered aqueous solution

concentration

~2 mg/mL

technique(s)

indirect immunofluorescence: 1-10 μg/mL

isotype

IgG1κ

fluorescence

λex 490 nm; λem 515 nm

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

6X His is a histidine tag that consists of a series of histidine residues at the amino or carboxy terminus of a protein.

Specificity

Binds 6X-His epitope tag

Immunogen

proteins contatining the epitope tag sequence

Application

Monoclonal Anti-6X His IgG, CF 488A antibody has been used in western blotting.

Biochem/physiol Actions

6X His tag does not affect the structure and function of protein and can be used in antibody production.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 1 mg/mL bovine serum albumin.

Preparation Note

Protect from light.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vernalization requirement duration in winter wheat is controlled by TaVRN?A 1 at the protein level.
Li G, et al.
The Plant Journal, 76(5), 742-753 (2013)
Haiyan Jia et al.
Plant physiology, 186(1), 483-496 (2021-02-13)
Programmed cell death (PCD) and apoptosis have key functions in development and disease resistance in diverse organisms; however, the induction of necrosis remains poorly understood. Here, we identified a semi-dominant mutant allele that causes the necrotic death of the entire
Genqiao Li et al.
The Plant journal : for cell and molecular biology, 76(5), 742-753 (2013-09-17)
Winter wheat requires a period of low temperatures to accelerate flowering (vernalization). This requirement could make winter wheat more vulnerable to elevated global temperature via insufficient vernalization. All known vernalization genes are cloned according to qualitative variation in vernalization requirement
Cătălin Voiniciuc et al.
Plant physiology, 169(1), 403-420 (2015-07-30)
Plants invest a lot of their resources into the production of an extracellular matrix built of polysaccharides. While the composition of the cell wall is relatively well characterized, the functions of the individual polymers and the enzymes that catalyze their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service