Skip to Content
Merck
All Photos(5)

Key Documents

SAB4503821

Sigma-Aldrich

Anti-phospho-HSL (pSer855/554) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,918.00

R$2,918.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$2,918.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,918.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 116 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer855/pSer554)

Gene Information

human ... LIPE(3991)

General description

Hormone sensitive lipase (HSL) is coded by LIPE (lipase E) gene. The gene coding for a protein is mapped on human chromosome 19q13. The encoded protein is an intracellular lipase with various neutral lipid substrates, such as triglycerides, diglycerides, monoglycerides, cholesterol esters and retinyl esters. HSL is widely expressed.

Immunogen

The antiserum was produced against synthesized peptide derived from human HSL around the phosphorylation site of Ser855/554.

Immunogen Range: 520-569

Biochem/physiol Actions

Hormone sensitive lipase (HSL) plays an important role in various processes such as lipolysis, adipocytes, steroidogenesis and spermatogenesis. Deficiency of HSL contributes to histologic abnormalities in white adipose tissue and an increase in macrophage infiltration. Polymorphism in the gene has been observed in familial lipodystrophies patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Null Mutation in Hormone-Sensitive Lipase Gene and Risk of Type 2 Diabetes
Albert JS, et al.
The New England Journal of Medicine, 370, 2307-2315 (2014)
A novel LIPE nonsense mutation found using exome sequencing in siblings with late-onset familial partial lipodystrophy.
Farhan SM, et al.
The Canadian Journal of Cardiology, 30, 1649-1654 (2014)
Identification of Microdeletions Spanning the Diamond-Blackfan Anemia Locus on 19q13 and Evidence for Genetic Heterogeneity
Gustavsson P, et al.
American Journal of Human Genetics, 63, 1388-1395 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service