Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502903

Sigma-Aldrich

Anti-Synapsin1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Brain protein 4.1, SYN-1, synapsin I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,950.00

R$3,950.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,950.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,950.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 73 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYN1(6853)

General description

Anti-Synapsin1 Antibody detects endogenous levels of total Synapsin1 protein.
Synapsin I (SYN1) is a synaptic vesicle associated protein. It is present in the central nervous system like olfactory bulb, cerebral cortex, hippocampus, basal ganglia, thalamus, hypothalamus, cerebellum, pons-medulla, spinal cord and posterior pituitary gland. SYN1 is found in the nucleus and cytoplasm of granule cells, and in the glomerular synaptic complex of the cerebellum. It is located in human chromosome Xp11.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human Synapsin1.

Immunogen Range: 26-75

Biochem/physiol Actions

Synapsin I (SYN1) plays an important role in neurotransmitter release, exocytosis of the synaptic vesicles, neurite elongation and synaptogenesis. Brain dysfunction occurs due to improper expression of synapsin I.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for a Turner syndrome locus or loci at Xp11. 2-p22. 1.
Zinn AR, et al.
American Journal of Human Genetics, 63(6), 1757-1766 (1998)
Increased synapsin I expression in cerebral malaria.
Thonsranoi K, et al.
International Journal of Clinical and Experimental Pathology, 8(11), 13996?14004-13996?14004 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service