Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500094

Sigma-Aldrich

Anti-YAP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

65 kDa Yes-associated protein, YAP1, YAP65

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,950.00

R$3,950.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,950.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,950.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YAP1(10413)

Related Categories

General description

Anti-YAP Antibody detects endogenous levels of total YAP protein.
Yes associated protein (YAP) is a 65kDa proline-rich phosphoprotein. It is encoded by the gene mapped to human chromosome 11q22.1. The encoded protein is characterized with a WW domain, a PDZ interaction motif, an SH3 binding motif and a coiled-coil domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human YAP.

Immunogen Range: 93-142

Biochem/physiol Actions

Yes associated protein (YAP) serves as a potent mechanosensitive transcriptional activator. It plays a vital role in regulation of cancer, regeneration and organ size. Overexpression of the gene contributes to tumorigenesis in breast and liver cancers. In addition, it also promotes gallbladder cancer (GBC) cell growth via activating AXL receptor tyrosine kinase/mitogen-activated protein kinase (MAPK) pathway. Thus, it acts as a potent biomarker and therapeutic target for various cancers including, GBC and breast cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yes-Associated Protein 1 Exhibits Oncogenic Property in Gastric Cancer and Its Nuclear Accumulation Associates with Poor Prognosis
Kang W, et al.
Clinical Cancer Research, 17(8), 2130-2139 (2011)
Force Triggers YAP Nuclear Entry by Regulating Transport across Nuclear Pores.
Elosegui-Artola A, et al.
Cell, 171(6), 1397-1410 (2017)
Expression of YES-associated protein (YAP) and its clinical significance in breast cancer tissues.
Cao L, et al.
Human Pathology, 68, 166-174 (2017)
Maolan Li et al.
Cancer letters, 355(2), 201-209 (2014-09-15)
The transcriptional coactivator Yes-associated protein 1 (YAP1), a key regulator of cell proliferation and organ size in vertebrates, has been implicated in various malignancies. However, little is known about the expression and biological function of YAP1 in human gallbladder cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service