Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301475

Sigma-Aldrich

Anti-phospho-Calmodulin (pThr79/Ser81) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CALM, CALM1, CALM2, CALM3, CAM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,820.00

R$2,820.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$2,820.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,820.00


Estimated to ship onMay 28, 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

rat, mouse, human

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_005175.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr79)

Gene Information

human ... CALM3(808)

General description

Calmodulin 3 (CALM3) encodes calmodulin (CaM) protein 3 which is expressed five folds more than CALM1 and CALM2. CALM3 promoter has high GC content and no TATA box like that of housekeeping gene. CALM3 contains several pro-alpha1 (AP1) and pro-alpha2 (AP2) consensus sites and several CAAT regions. CALM3 also has clusters of specificity protein 1 (SP1) transcription factor. In human chromosome, the gene is localized on 19q13.32.

Specificity

The antibody detects endogenous level of Calmodulin only when phosphorylated at threonine 79/serine 81.

Immunogen

Peptide sequence around phosphorylation site of threonine79/serine81(K-D-T(p)-D-S(p)-E-E) derived from Human Calmodulin .

Biochem/physiol Actions

Calmodulin (CaM) encoded by Calmodulin 3 (CALM3) senses Ca2+ and binds to free signal-transducing protein that regulates Cav1.2 L-type calcium channel and deactivates Cav1.2 when CaM binds to free Ca2+. A sporadic mutation in CALM3 might be the cause for long QT syndrome, a disorder of ventricular myocardial repolarization, characterized by prolonged heart rate. CALM3 expression is higher in teratoma cells. CALM3 is a potential biomarker to dysregulate the melanoma cancer genes. Polymorphism in CALM3 is a potential modifier gene for familial hypertrophic cardiomyopathy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumour-suppressive microRNA-224 inhibits cancer cell migration and invasion via targeting oncogenic TPD52 in prostate cancer
Goto Y, et al.
Febs Letters, 588(10), 1973-1982 (2014)
Characterization of the human CALM2 calmodulin gene and comparison of the transcriptional activity of CALM1, CALM2 and CALMS
Toutenhoofd SL, et al.
Cell Calcium, 23(5), 323-338 (1998)
Identification of melanoma biomarkers based on network modules by integrating the human signalling network with microarrays
Huang C, et al.
Journal of Cancer Research and Therapeutics, 10(7), 114-124 (2014)
A new polymorphism in human calmodulin III gene promoter is a potential modifier gene for familial hypertrophic cardiomyopathy
Friedrich FW, et al.
European Heart Journal, 30(13), 1648-1655 (2009)
CALM3 mutation associated with long QT syndrome
Reed GJ, et al.
Heart Rhythm : the Official Journal of the Heart Rhythm Society, 12(2), 419-422 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service