Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300685

Sigma-Aldrich

Anti-ASH2L antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ASH2, Anti-ASH2L1, Anti-ASH2L2, Anti-Bre2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,459.00

R$2,459.00


Estimated to ship onMay 24, 2025



Select a Size

Change View
100 μG
R$2,459.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,459.00


Estimated to ship onMay 24, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65-80 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(G-R-R-S-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASH2L(9070)

General description

The antibody detects endogenous level of total ASH2L protein.

Immunogen

Peptide sequence around aa. 620-624 (G-R-R-S-P), according to the protein NP_004665.2

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Component of the Set1/Ash2 histone methyltransferase (HMT) complex, a complex that specifically methylates ′Lys-4′ of histone H3, but not if the neighboring ′Lys-9′ residue is already methylated. As part of the MLL1/MLL complex it is involved in methylation and dimethylation at ′Lys-4′ of histone H3. May function as a transcriptional regulator. May play a role in hematopoiesis.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service