Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300561

Sigma-Aldrich

Anti-SMAD1 (Ab-465) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSP1 antibody produced in rabbit, Anti-JV4-1 antibody produced in rabbit, Anti-JV41 antibody produced in rabbit, Anti-MADH1 antibody produced in rabbit, Anti-SMAD family member 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,443.00

R$3,443.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μG
R$3,443.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,443.00


Estimated to ship onMay 28, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~55 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-S-S-V-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD1(4086)

General description

Smad1 (SMAD family member 1) is a transcriptional activator, that is located on chromosome 4q31.

Immunogen

Peptide sequence around aa. 461-465 (I-S-S-V-S), according to the protein SMAD1.

Biochem/physiol Actions

SMAD1 (SMAD family member 1) protein has a major role in disease development, hence it acts as a biologic prognostic factor in diseases. The miR-345 repress the multiplication, invasion and migration of prostate cancer by downregulating Smad1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD1 is a receptor-regulated SMAD (R-SMAD).

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The distribution and potential prognostic value of SMAD protein expression in chronic lymphocytic leukemia
Witkowska M, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(3) (2017)
An interstitial 4q31.21q31.22 microdeletion associated with developmental delay: case report and literature review
Vlaikou AM, et al.
Cytogenetic and genome research, 142(4), 227-238 (2014)
Integrating patterning signals: Wnt/GSK3 regulates the duration of the BMP/Smad1 signal
Fuentealba LC, et al.
Cell, 131(5), 980-993 (2007)
MiR-345 suppresses proliferation, migration and invasion by targeting Smad1 in human prostate cancer
Chen QG, et al.
Journal of Cancer Research and Clinical Oncology, 142(1), 213-224 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service