Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300438

Sigma-Aldrich

Anti-ZAP70 (Ab-493) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ17670 antibody produced in rabbit, Anti-FLJ17679 antibody produced in rabbit, Anti-SRK antibody produced in rabbit, Anti-STD antibody produced in rabbit, Anti-zeta-chain (TCR) associated protein kinase 70kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,582.00

R$2,582.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$2,582.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,582.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~70 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(S-Y-Y-T-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZAP70(7535)

Immunogen

Peptide sequence around aa. 491-495 (S-Y-Y-T-A), according to the protein ZAP70.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

ZAP70 is a 70-kD tyrosine phosphoprotein that associates with the zeta chain and undergoes tyrosine phosphorylation following TCR stimulation. The ZAP70 gene is expressed in T- and natural KILLER cells. Protein-Tyrosine Kinases (PTKs) play an integral role in T-cell activation. Stimulation of the T-cell antigen receptor results in tyrosine phosphorylation of a number of cellular substrates. One of these is the TCR-zeta chain, which can mediate the transduction of extracellular stimuli into cellular effector functions.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service