Skip to Content
Merck
All Photos(4)

Key Documents

SAB4300427

Sigma-Aldrich

Anti-PAK1 (Ab-212) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC130000 antibody produced in rabbit, Anti-MGC130001 antibody produced in rabbit, Anti-PAKalpha antibody produced in rabbit, Anti-p21 protein (Cdc42 / Rac)-activated kinase 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~68 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-V-T-P-T)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAK1(5058)

Immunogen

Peptide sequence around aa. 210-214 (P-V-T-P-T), according to the protein PAK1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The activated kinase acts on a variety of targets. Likely to be the GTPase effector that links the Rho-related GTPases to the JNK MAP kinase pathway. Activated by CDC42 and RAC1. Involved in dissolution of stress fibers and reorganization of focal complexes. Involved in regulation of microtubule biogenesis through phosphorylation of TBCB. Activity is inhibited in cells undergoing apoptosis, potentially due to binding of CDC2L1 and CDC2L2.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Rettig et al.
PloS one, 7(7), e42012-e42012 (2012-08-01)
The role of c-Crk (CRK) in promoting metastasis is well described however the role of CRK phosphorylation and the corresponding signaling events are not well explained. We have observed CRK-II serine 41 phosphorylation is inversely correlated with p120-catenin and E-cadherin
Fariborz Mortazavi et al.
BMC cancer, 15, 381-381 (2015-05-10)
Key effector(s) of mutated KRAS in lung cancer progression and metastasis are unknown. Here we investigated the role of PAK1/Crk axis in transduction of the oncogenic KRAS signal in non-small cell lung cancer (NSCLC). We used NSCLC clinical specimens to
Gholam H Pezeshkpour et al.
Genes & cancer, 4(7-8), 315-324 (2013-10-30)
CRK (c-Crk) as an adaptor protein is involved in several oncogenic signal transduction pathways, conveying oncogenic signals to its downstream effectors and thereby affecting multiple cellular processes including proliferation, differentiation, and migration. For example, we have observed that CRK expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service