Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300117

Sigma-Aldrich

Anti-phospho-VAV1 (pTyr174) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-VAV antibody produced in rabbit, Anti-vav 1 guanine nucleotide exchange factor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,279.00

R$3,279.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
100 μG
R$3,279.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,279.00


Estimated to ship onJune 02, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~95 kDa

species reactivity

rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-I-YP-E-D)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr174)

Gene Information

human ... VAV1(7409)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 174 (E-I-Y(p)-E-D), according to the protein VAV1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Couples tyrosine kinase signals with the activation of the Rho/Rac GTPases, thus leading to cell differentiation and/or proliferation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service