Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200842

Sigma-Aldrich

Anti-PMEL antibody, Mouse monoclonal

clone PL-19, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$833.00
100 μL
R$2,197.00

R$833.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$833.00
100 μL
R$2,197.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$833.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

PL-19

form

liquid

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using human melanoma SK-MEL-28 cell lysate
immunofluorescence: 1-2 μg/mL using human melanoma SK-MEL-28 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PMEL(6490)

General description

The melanocyte-specific protein PMEL (also known as Pmel17, gp100, melanocyte protein, premelanosome protein or silver locus protein homolog (SILV) is a type I transmembrane glycoprotein that is expressed primarily in pigment cells of the eye and skin. In melanosomes, PMEL protein forms a fibrillar matrix on which the UV-shielding pigment melanin is deposited.1-2

Specificity

Monoclonal Anti-PMEL antibody specifically recognizes human PMEL protein.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (~100 kDa), Immunohistochemistry and Immunofluorescence.

Biochem/physiol Actions

PMEL fibrils are a major functional component of the melanosomal compartment as they optimize the melanin polymerization, condensation and storage.1,3
In addition, PMEL fibrils have an amyloidogenic nature sharing features with pathological amyloids.4 Thus, PMEL is suggested as an excellent model system to study mechanisms of intracellular amyloid formation.1
Mutations in PMEL are associated with pigmentation disorders and/or impairments in eye development in various species.1,5,6

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service