Skip to Content
Merck
All Photos(3)

Documents

SAB4200828

Sigma-Aldrich

Anti-Pseudomonas Exotoxin A-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

ETA, NAD(+)--diphthamide ADP-ribosyltransferase, PE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

lyophilized powder

species reactivity

Pseudomonas aeruginosa

concentration

~4 mg/mL

technique(s)

direct ELISA: 1:10,000-1:20,000 (is recommended using 1 μg/ml Pseudomonas Exotoxin A for coating.)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Pseudomonas aeruginosa PAO1 ... toxA(877850)

General description

Pseudomonas aeruginosa is a Gram negative bacterium that rarely cause diseases in humans but certain populations, such as cystic fibrosis (CF) or burn wounds patients, are prone to such infections.
Pseudomonas Exotoxin A (PE) is the most potent virulence factor secreted by P. aeruginosa, it is composed of three structural domains, N-terminal domain (I) responsible for the toxin binding to its host cell receptor, middle domain (II) has a role in toxin translocation across the membrane, and C-terminal domain (III) has ADP-ribosylation activity. PE is considered as a selective agent for the elimination of specific cell populations due to its toxin ADP-ribosylation activity resulting in irreversibly shut down of protein synthesis. To reduce the adverse effects of natural PE, Mutated PE was used in several attempts to develop recombinant toxin-antibody or cytokines fusion fragments for therapeutic application including cancer immunotherapy.

Immunogen

Purified Exotoxin A from Pseudomonas aeruginosa

Application

The antibody may be used in various immunochemical techniques including ELISA.

Physical form

Supplied as a lyophilized powder.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service