Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200816

Sigma-Aldrich

Anti-Calcineurin (α-Subunit) antibody, Mouse monoclonal

clone CN-A1, purified from hybridoma cell culture

Synonym(s):

CAM-PRP catalytic subunit, Calmodulin-dependent calcineurin A subunit alpha isoform, Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,125.00
100 μL
R$3,661.00

R$1,125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,125.00
100 μL
R$3,661.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,125.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

CN-A1, monoclonal

species reactivity

bovine, human, rat

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1.25-2.5 μg/mL using mouse or rat brain tissue extract.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... PPP3CA(286852)
human ... CALNA(5530)
rat ... Caln1(363909)

Related Categories

General description

Calcineurin (a-subunit) (CnA), also known as Serine/threonine-protein phosphatase 2B catalytic subunit alpha (PP3CA), Calmodulin-dependent calcineurin A subunit alpha (CALN/CALNA) or CAM-PRP catalytic subunit, is a major soluble calmodulin-binding protein in the brain. Calcineurin is a heterodimer metalloenzyme, Ca2+/Calmodulin dependent serine/threonine protein phosphatase 3 (PPP3), with a relatively narrow substrate specificity4-6,8 It is composed of two subunits, a catalytic calmodulin-binding a subunit (61 kDa, calcineurin A [CnA]) and regulatory calcium-binding ß subunit (18 kDa, calcineurin B [CnB]).6,7 Both subunits are highly conserved from yeast to mammals.8

Specificity

Monoclonal Anti-Calcineurin specifically recognizes an epitope located on the a-subunit of calcineurin from human, mouse, monkey, porcine1, rat2 and bovine13 origin. It does not cross-react with calcineurin ß-subunit.

Immunogen

Purified bovine brain calcineurin.

Application

The antibody may be used in various immunochemical techniques including Immunoblot1,2 (~61 kDa), ELISA and Immunohistochemistry1,3.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Niitsu et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 83(6), 308-315 (2016-06-21)
Increased expression of calcineurin in colorectal cancer (CRC) has been reported. Although the oncogenic function has been suggested, the clinical relevance is still unclear. We herein studied calcineurin expression as a prognostic biomarker in patients receiving curative surgery for stages

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service