Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200795

Sigma-Aldrich

Anti-Exendin-4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Exenatide

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,876.00

R$2,876.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
100 μL
R$2,876.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,876.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

concentration

~1 mg/mL

technique(s)

indirect ELISA: 0.125-0.25 μg/mL using 1 μg/mL Exendin-4 protein for coating

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Exendin-4 is an incretin mimetic 39-amino-acid peptide found in venom from the Gila monster (Helicoderma suspectum). Exendin-4 belongs to the glucagon-secretin family of peptide hormones and neuropeptides. It acts as a potent agonist of the GLP-1 receptor and is an insulinotropic agent (promotes insulin secretion) with a long half-life.

Exendin-4 mimics the activity of mammalian incretin hormone glucagon-like peptide 1 (GLP-1) and thus play a role in the control of glucose. This includes the secretion of insulin in a glucose-dependent manner, negative regulation of high glucagon secretion, and increased duration of stomach emptying. In type 2 diabetes patients, this peptide can be administered subcutaneously for glycemic control, when metformin is unable to produce adequate results. Exendin-4 promotes the neogeneration and proliferation of β-cells, and thus aids in the regeneration of pancreas. It acts as a ligand to exendin receptor and leads to an elevation of acinar cell cAMP levels.

Anti-Exendin-4 antibody specifically recognizes synthetic Exendin-4 and does not cross react with GLP-1.

Immunogen

synthetic peptide corresponding to the C-terminal sequence of Heloderma suspectum (Gila monster) Exendin-4 , conjugated to KLH

Application

The antibody may be used in various immunochemical techniques including ELISA

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service