Skip to Content
Merck
All Photos(3)

Documents

SAB4200652

Sigma-Aldrich

Monoclonal Anti-TOM1L1 antibody produced in mouse

clone 3F12, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~53 kDa

species reactivity

human, bovine, monkey

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.5-1 μg/mL using using whole extracts of HeLa cells.
immunofluorescence: 5-10 μg/mL using using HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOM1L1(10040)

General description

The gene TOM1L1 (target of myb1 like 1 membrane trafficking) is mapped to human chromosome 17q23. The protein has a VHS (Vps27p/Hrs/Stam) domain at the N-terminal, a GAT (GGA (Golgi-localizing, γ-adaptin ear domain homology, ADP-ribosylation factor (Arf)-binding protein and Tom) domain at the center and interaction motifs at the C-terminal.

Immunogen

immunized with a TOM1L1 fusion protein.

Biochem/physiol Actions

TOM1L1 (target of myb1 like 1 membrane trafficking) is an adaptor protein, which is involved in vesicular trafficking as well as intracellular signaling. It negatively controls the mitogenic and transforming functions in fibroblasts by working as a substrate for SRC (tyrosine protein kinase). It is involved in EGF (epidermal growth factor)-mediated internalization of EGF receptor. TOM1L1 is downregulated in cutaneous squamous cell carcinoma and esophageal squamous cell carcinoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Béatrice Orsetti et al.
Cancer research, 64(18), 6453-6460 (2004-09-18)
Chromosome 17 is severely rearranged in breast cancer. Whereas the short arm undergoes frequent losses, the long arm harbors complex combinations of gains and losses. In this work we present a comprehensive study of quantitative anomalies at chromosome 17 by
Yu Qi et al.
Anticancer research, 30(9), 3535-3539 (2010-10-15)
Src-family tyrosine kinases (SFKs) play critical roles in regulating cellular differentiation and proliferation. Src-activating and signaling molecule (Srcasm) is a novel molecule that down-regulates SFK activity and promotes cell differentiation. The aim of this study was to determine whether Srcasm
Ning Sheng Liu et al.
The EMBO journal, 28(22), 3485-3499 (2009-10-03)
Although many proteins have been shown to participate in ligand-stimulated endocytosis of EGF receptor (EGFR), the adaptor protein responsible for interaction of activated EGFR with endocytic machinery remains elusive. We show here that EGF stimulates transient tyrosine phosphorylation of Tom1L1
Ahmed Elmarghani et al.
Mediators of inflammation, 2009, 416298-416298 (2010-02-26)
TOM1L (target of Myb-1 Like) was identified as a binding partner for the full length and catalytically-active Lck in a yeast 2-hybrid screening assay. Here we show that in Jurkat T cells stimulated by CD3/CD28 coligation where the expression of
Audrey Sirvent et al.
Molecular & cellular proteomics : MCP, 11(12), 1937-1950 (2012-10-02)
The non-receptor tyrosine kinase SRC is frequently deregulated in human colorectal cancer (CRC), and SRC increased activity has been associated with poor clinical outcomes. In nude mice engrafted with human CRC cells, SRC over-expression favors tumor growth and is accompanied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service