Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200612

Sigma-Aldrich

Monoclonal Anti-EXOC6 antibody produced in mouse

~1.0 mg/mL, clone 15S2G6, purified immunoglobulin

Synonym(s):

Anti-EXOC6A, Anti-RP11-348J12.4, Anti-SEC15-like 1, Anti-SEC15-like protein 1, Anti-SEC15-like protein 3, Anti-SEC15, Anti-SEC15L1, Anti-SEC15L3, Anti-SEC15L, Anti-Sec15p, Anti-exocyst complex component Sec15A

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
R$2,709.00

R$2,709.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
R$2,709.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,709.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15S2G6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

canine, rat, monkey, bovine, human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 20 μg/test using A549 cells
immunoblotting: 0.5-1.0 μg/mL using NT2/D1 total cell extracts
immunofluorescence: 20 μg/mL using PC12 or A549 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... EXOC6(50556)

Related Categories

General description

Monoclonal Anti-EXOC6 (mouse IgG1 isotype) is derived from the hybridoma 15S2G6 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant protein corresponding to full-length rat brain EXOC6 subunit. Exocyst complex component 6 (EXOC6) is located on the human chromosome at 10q23.33.

Immunogen

a recombinant protein corresponding to full-length rat brain EXOC6 subunit.

Application

Monoclonal Anti-EXOC6 antibody has been used in
  • immunoblotting
  • immunofluorescence
  • flow Cytometry
  • immunocytochemistry

Biochem/physiol Actions

Exocyst complex component 6 (EXOC6) has been shown to interact with the Rab11 (Ras-related protein), a GTPase that regulates both biosynthetic and endocytic traffic. This interaction results in endocytic trafficking directed toward the apical and basolateral poles of polarized cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide gene-environment interaction analysis for asbestos exposure in lung cancer susceptibility
Wei S, et al.
Carcinogenesis, 33(8), 1531-1537 (2012)
Rab proteins and the compartmentalization of the endosomal system
Wandinger-Ness A and Zerial M
Cold Spring Harbor Perspectives in Biology, 6(11), a022616-a022616 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service