Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200337

Sigma-Aldrich

Anti-phospho-RasGRP1(pThr184) antibody, Mouse monoclonal

clone p-RasGRP1-1, purified from hybridoma cell culture

Synonym(s):

Anti-CALDAG-GEFI, Anti-CALDAG-GEFII, Anti-RAS guanyl releasing protein 1 (calcium and DAG-regulated) hRasGRP1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
R$4,044.00

R$4,044.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
R$4,044.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,044.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

p-RasGRP1-1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 2-4 μg/mL using whole extracts of HEK-293T cells overexpressing human RasGRP1

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr184)

Gene Information

General description

Monoclonal Anti-phospho-RasGRP1(pThr184) (mouse IgG2a isotype) is derived from the hybridoma p-RasGRP1-1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic phospho-peptide corresponding to residues surrounding Thr184 of human RasGRP1 conjugated to KLH. RAS guanyl releasing protein 1 (RasGRP1) is a member of RasGRP family, characterized by the presence of a N-terminal Ras superfamily guanine nucleotide exchange factor (GEF) domain and and C-terminal calcium- and diacylglycerol (DAG)/phorbol ester-binding domains. It is an intracellular signaling protein. RasGRP1 has a N terminal guanine nucleotide exchange factor domain for Ras-like guanosine triphosphatases (GTPases) (RasGEFN/RasGEF), two EF hand motifs and a carboxy-terminal C1 domain.

Immunogen

a synthetic phospho-peptide corresponding to residues surrounding Thr184 of human RasGRP1, conjugated to KLH. The corresponding sequence is identical in rat, mouse, monkey, bovine and canine RasGRP1.

Application

Monoclonal Anti-phospho-RasGRP1(pThr184) antibody produced in mouse has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

RAS guanyl releasing protein 1 (RasGRP1) functions as a diacylglycerol (DAG)-regulated nucleotide exchange factor specifically activating Ras through the exchange of bound guanosine diphosphate (GDP) for guanosine triphosphate (GTP). It activates the extracellular signal-regulated kinase (Erk)/mitogen-activated protein (MAP) kinase cascade and regulates T-cells and B-cells development, homeostasis and differentiation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galphai2 and ZAP-70 mediate RasGRP1 membrane localization and activation of SDF-1-induced T cell functions
Kremer KN, et al.
Journal of Immunology, 187(6), 3177-3185 (2011)
Defective expression of Ras guanyl nucleotide-releasing protein 1 in a subset of patients with systemic lupus erythematosus
Yasuda S, et al.
Journal of Immunology, 179(7), 4890-4900 (2007)
Divergent and convergent signaling by the diacylglycerol second messenger pathway in mammals
Brose N, et al.
Current Opinion in Neurobiology, 14(3), 328-340 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service