SAB4200065
Anti-EIF2C1/Ago1 antibody produced in rabbit
~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Argonaute-1, Anti-Eukaryotic translation initiation factor 2C, subunit 1, Anti-GERP95, Anti-Golgi Endoplasmic Reticulum protein 95 kDa
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~100 kDa
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): 5-10 μg using lysates of HEK-293T cells over expressing hEIF2C1/Ago1
indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing hEIF2C1/Ago1
western blot: 2-4 μg/mL using lysates of HEK-293T cells over expressing hEIF2C1/Ago1
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... EIF2C1(26523)
mouse ... Eif2c1(236511)
rat ... Eif2c1(313594)
Application
Western Blotting (1 paper)
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service