Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701015

Sigma-Aldrich

Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

HRP

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
R$1,801.00

R$1,801.00


Estimated to ship onApril 29, 2025



Select a Size

Change View
500 μG
R$1,801.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

R$1,801.00


Estimated to ship onApril 29, 2025


biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Peroxidase, Anti-Goat Serum, Mouse IgG, Mouse IgG F(ab′)2 and Mouse Serum. No reaction was observed against Anti-Pepsin, Anti-Goat IgG F(c), Mouse IgG F(c) or Bovine, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.

Immunogen

Mouse IgG F(ab′)2 fragment

Physical properties

Antibody format: IgG F(ab′)2

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 500 μ;L deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José-Daniel Aroca-Aguilar et al.
PloS one, 13(12), e0209364-e0209364 (2018-12-18)
Myocilin is an extracellular glycoprotein with a poorly understood biological function and typically known because of its association with glaucoma. In this study, we analyzed the expression and biological activity of human myocilin in some non-ocular tissues. Western immunoblot showed
Tomoshige Wakayama et al.
Asian Pacific journal of allergy and immunology (2021-09-21)
The role of anti-elastin antibody (Ab) in the lung is unclear, although they may be involved in chronic obstructive pulmonary disease (COPD). Recently, increased anti-elastin Ab levels were reported in asthma. To elucidate the role of anti-elastin Ab in asthma
Jessica L Ray et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 38(1), e23350-e23350 (2023-12-10)
Lung diseases characterized by type 2 inflammation are reported to occur with a female bias in prevalence/severity in both humans and mice. This includes previous work examining multi-walled carbon nanotube (MWCNT)-induced eosinophilic inflammation, in which a more exaggerated M2a phenotype

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service