Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501167

Sigma-Aldrich

Monoclonal Anti-Seasonal H1N1 Neuraminidase [10C5F12] antibody produced in mouse

purified antibody (Protein A)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,255.00

R$3,255.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,255.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,255.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody (Protein A)

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

Influenza A virus subtype H1N1

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Seasonal H1N1 Neuraminidase Monoclonal Antibody: Influenza A virus has one of sixteen possible hemagglutinin (HA) surface proteins and one of nine possible neuraminidase (NA) surface proteins. In early 2009, a novel H1N1 swine-origin influenza (S-OIV) A virus was identified in specimens obtained from patients in Mexico and the United States. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin.

Immunogen

Mouse monoclonal neuraminidase antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.

Application

Neuraminidase antibody can be used for the detection of neuraminidase protein from seasonal influenza A (H1N1) in ELISA. It will also detect 50 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service