Skip to Content
Merck
All Photos(3)

Key Documents

SAB3501035

Sigma-Aldrich

Anti-KREMEN1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,623.00

R$2,623.00


Estimated to ship onMay 07, 2025



Select a Size

Change View
100 μG
R$2,623.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,623.00


Estimated to ship onMay 07, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 54 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KREMEN1(83999)

Immunogen

Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KREMEN1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501035.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke Wang et al.
The Journal of biological chemistry, 283(34), 23371-23375 (2008-05-27)
Wnt signaling is involved in a wide range of developmental, physiological, and pathophysiological processes and is negatively regulated by Dickkopf1 (Dkk1). Dkk1 has been shown to bind to two transmembrane proteins, the low density lipoprotein receptor-related proteins (LRP) 5/6 and
Jing Li et al.
Fertility and sterility, 90(4 Suppl), 1470-1479 (2007-12-11)
To determine the roles of Dickkopf-1 (Dkk1) in mouse embryo implantation. Experimental prospective study. Animal research and institute laboratory facility. Virgin Kunming female mice and adult male mice. The expression of Dkk1 and its receptor Kremen1 in embryos and uteri
Bingyu Mao et al.
Nature, 417(6889), 664-667 (2002-06-07)
The Wnt family of secreted glycoproteins mediate cell cell interactions during cell growth and differentiation in both embryos and adults. Canonical Wnt signalling by way of the beta-catenin pathway is transduced by two receptor families. Frizzled proteins and lipoprotein-receptor-related proteins
T Nakamura et al.
Biochimica et biophysica acta, 1518(1-2), 63-72 (2001-03-27)
Kringle domain, a triple-disulfide-linked domain, is conserved in diverse proteins which play important roles in various biological processes. We cloned Kremen, a novel member of kringle-containing proteins, using a newly developed unique strategy, 'Kringle-SAGE (serial analysis of gene expression)', which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service