Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500471

Sigma-Aldrich

Monoclonal Anti-EndoG antibody produced in mouse

clone 7F2G10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Endonuclease G

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,092.00

R$3,092.00


Estimated to ship onMay 02, 2025



Select a Size

Change View
100 μG
R$3,092.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,092.00


Estimated to ship onMay 02, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F2G10, monoclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENDOG(2021)

General description

ENDOG (Endonuclease G) is a 28kDa mitochondria-localized protein mapped to human chromosome 9q34.1. However, in presence of certain conditions it can translocate to the nucleus through the BNIP3 (Bcl-2/adenovirus E1B 19kDa protein-interacting protein 3) cell death pathway.

Immunogen

Mouse monoclonal EndoG antibody was raised against a recombinant protein corresponding to amino acids 51 – 140 of human EndoG.

Application

Monoclonal Anti-EndoG antibody produced in mouse is suitable for indirect ELISA and western blot.

Biochem/physiol Actions

ENDOG (Endonuclease G) is a mitochondrial apoptotic DNase involved in the BNIP3 (Bcl-2/adenovirus E1B 19kDa protein-interacting protein 3) cell death pathway. During mitochondrial release and nuclear translocation of EndoG, BNIP3 interacts with the voltage-dependent anion channel (VDAC) to facilitate the process in the nucleus, it cleaves chromatin apoptotic DNA without the need of caspase activity. ENDOG causes DNA fragmentation during and after apoptosis. It is linked to cardiac hypertrophy and Parkinson′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaosha Zhang et al.
PloS one, 9(12), e113642-e113642 (2014-12-02)
BNIP3 is a proapoptotic protein that induces cell death through a mitochondria-mediated pathway. We reported previously that mitochondrial localization of BNIP3 and translocation of EndoG from mitochondria to the nucleus are critical steps of the BNIP3 pathway. It is not
Dae Song Jang et al.
DNA and cell biology, 34(2), 92-100 (2014-11-18)
Apoptotic endonuclease G (EndoG) is responsible for DNA fragmentation both during and after cell death. Previous studies demonstrated that genetic inactivation of EndoG is cytoprotective against various pro-apoptotic stimuli; however, specific inhibitors for EndoG are not available. In this study
V Tiranti et al.
Genomics, 25(2), 559-564 (1995-01-20)
By using a PCR-based screening of a somatic cell hybrid panel and FISH, we have assigned the loci of mitochondrial single-stranded DNA-binding protein (SSBP), mitochondrial transcription factor A (TCF6), and mitochondrial endonuclease G (ENDOG) genes to human chromosomes 7q34, 10q21

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service