Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500124

Sigma-Aldrich

Monoclonal Anti-CD4L antibody produced in mouse

clone 9H5A8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD4 antigen, Anti-CD4 receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,731.00

R$3,731.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1759


Select a Size

Change View
100 μG
R$3,731.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,731.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1759

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H5A8, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD4(920)

General description

Cluster of differentiation (CD4) encodes a T-cell surface glycoprotein CD4. In human chromosome, the gene CD4 is localized on 12p13.31.

Immunogen

raised against a 193 amino acid recombinant protein from near the amino terminus of human CD4.

Biochem/physiol Actions

Cluster of differentiation (CD4) mutation in single nucleotide causes OKT4 (Leu-3 T4) epitope deficiency. CD4 plays a major role in collateral artery formation by recruiting macrophages at the active site and through triggering the synthesis of arteriogenic cytokines.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

CD4 is a member of the immunoglobulin superfamily and is implicated as associative recognition element in MHC (Major Histocompatibility Complex) class II-restricted immune response. On T-lymphocytes, it defines the helper/inducer subset. The mature 55 kd CD4 protein consists of a 372 amino acid extracellular segment composed of four tandem immunoglobulin-like VJ regions. The CD4 molecule is a major receptor for human immunodeficiency virus (HIV), binding directly to the envelope glycoprotein gp120 on HIV, with the co-receptors being CCR5 or CXCR4. It has been shown that the V-like domains are critical for binding with HIV envelope gp120.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene-expression profiling of titanium-cell interaction
Kim CS, et al.
The Journal of Korean Academy of Prosthodontics, 43(3), 393-408 (2005)
OKT4 epitope deficiency in patients infected with the human immunodeficiency virus: a cause of underestimation of the CD4 lymphocyte count
Khuong MA, et al.
Clinical Infectious Diseases, 19(5), 961-963 (1994)
Impaired Arteriogenic Response to Acute Hindlimb Ischemia in CD4-Knockout Mice
Stabile E, et al.
Cytokines, 108(5), 205-210 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service