Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500070

Sigma-Aldrich

Anti-THEM2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ACOT13, Anti-Acyl-CoA thioesterase 13, Anti-PNAS-27, Anti-Thioesterase superfamily member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,798.00

R$2,798.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$2,798.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,798.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACOT13(55856)

General description

Acyl-CoA thioesterase 13 (ACOT13), also known as thioesterase superfamily member 2 (THEM-2), is encoded by the gene mapped to human chromosome 6p22.3. The encoded protein has a molecular mass of ∼15kDa with 140 amino acids. It is a member of the hotdog-domain thioesterase family. THEM2 is highly expressed in several tissues such as heart, kidney, liver and brown adipose tissue (BAT). It has been shown to be co-localized with β-tubulin on microtubules. At least two isoforms of THEM2 are known to exist.

Immunogen

THEM2 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human THEM2.

Biochem/physiol Actions

Acyl-CoA thioesterase 13 (ACOT13)/thioesterase superfamily member 2 (THEM-2) possesses thioesterase activity, with greater activity observed with longer chain acyl-CoAs such as myristoyl- and palmitoyl-CoA. THEM2 associates with StarD, a protein that plays a key role in fatty acid metabolism, and the addition of StarD to THEM2 increased its catalytic activity, suggesting that THEM2 plays a significant role in the metabolism of fatty acids.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500070.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thioesterase superfamily member 2 (Them2)/acyl-CoA thioesterase 13 (Acot13): a homotetrameric hotdog fold thioesterase with selectivity for long-chain fatty acyl-CoAs.
Jie W, et al.
The Biochemical Journal, 421(2), 311-322 (2009)
Deactivating fatty acids: acyl-CoA thioesterase-mediated control of lipid metabolism.
Tillander V, et al.
Trends in Endocrinology and Metabolism, 28(7), 473-484 (2017)
Molecular features of the basal-like breast cancer subtype based on BRCA1 mutation status.
Prat A, et al.
Breast Cancer Research and Treatment, 147(1), 185-191 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service