Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900618

Sigma-Aldrich

Anti-HTR6/5-HT36 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

5-HT-6, 5-HT6, 5-HT6 receptor, 5-HT6R, 5-hydroxytryptamine receptor 6, HTR6

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$2,675.00

R$2,675.00


Check Cart for Availability


Select a Size

Change View
50 μG
R$2,675.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,675.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

horse, dog, rat, hamster, human, monkey

technique(s)

immunohistochemistry: 20 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTR6(3362)

Immunogen

Synthetic 17 amino acid peptide from C-terminus of human HTR6 / 5-HT36. Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Rat, Hamster, Panda, Dog, Horse (100%); Mouse, Elephant, Bovine, Bat, Rabbit (94%); Stickleback, Pufferfish (88%); Turkey, Chicken, Xenopus, Zebrafish (82%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.
Supplied at 1 mg/ml

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service