Skip to Content
Merck
All Photos(1)

Key Documents

SAB2702344

Sigma-Aldrich

Monoclonal Anti-Aspartate aminotransferase antibody produced in mouse

clone GT638, affinity isolated antibody

Synonym(s):

AST1, ASTQTL1, GIG18, cAspAT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,466.00

R$3,466.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
100 μL
R$3,466.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,466.00


Estimated to ship onJune 02, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT638, monoclonal

form

buffered aqueous solution

mol wt

46kDa

species reactivity

human, mouse

concentration

1mg/mL

technique(s)

western blot: 1:500-1:3000

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOT1(2805)

Immunogen

Recombinant protein encompassing a sequence within the N-terminus region of human Aspartate aminotransferase.

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein A

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service