Skip to Content
Merck
All Photos(4)

Key Documents

SAB2702134

Sigma-Aldrich

Monoclonal Anti-BRCA1 antibody produced in mouse

Synonym(s):

PNCA4, PPP1R53, PSCP, RNF53

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,508.00

R$2,508.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1675


Select a Size

Change View
100 μL
R$2,508.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,508.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1675

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

17F8
monoclonal

form

liquid

mol wt

220 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

ChIP: suitable
ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

Immunogen

Protein fragment expressed in E. coli corresponding to amino acids 762-1315.

Application

For ICC/IF: Use at a concentration of 1 μg/mL. For WB: 1:500-1:3000. For IHC-P: Use at an assay dependent dilution. Optimal dilutions/concentrations should be determined by the researcher

Biochem/physiol Actions

BRCA1-17F8 recognizes full-length BRCA1, a 220kDa nuclear phosphoprotein, and does not recognize the Aexon 11 splice variant. In a high proportion of breast and ovarian cancer cell lines, BRCA1 aberrantly mislocates to the cytoplasm. Its usefulness to monitor functional inactivation of BRCA1 in sporadic breast cancers is under active investigation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service