Skip to Content
Merck
All Photos(8)

Key Documents

SAB2701975

Sigma-Aldrich

Anti-KLF4 antibody produced in rabbit

Synonym(s):

EZF, GKLF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,508.00

R$2,508.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,508.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,508.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

51 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

flow cytometry: 50-200
immunoprecipitation (IP): suitable
western blot: 500-10000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KLF4(9314)

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 349 and 457 of KLF4

Application

Suggested starting dilutions are as follows: FACS: 1:50-1:200, IP: 1:100-1:500, WB: 1:500-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Transcription factor which acts as both an activator and repressor. Binds the CACCC core sequence. Binds to multiple sites in the 5′-flanking region of its own gene and can activate its own transcription. Required for establishing the barrier function of the skin and for postnatal maturation and maintenance of the ocular surface. Involved in the differentiation of epithelial cells and may also function in skeletal and kidney development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel S Kim et al.
Nature genetics, 53(11), 1564-1576 (2021-10-16)
Transcription factors bind DNA sequence motif vocabularies in cis-regulatory elements (CREs) to modulate chromatin state and gene expression during cell state transitions. A quantitative understanding of how motif lexicons influence dynamic regulatory activity has been elusive due to the combinatorial
Ming-Quan Ma et al.
International journal of clinical and experimental pathology, 7(10), 6679-6685 (2014-11-18)
To investigate the association of Kruppel-like factor 4 (KLF4) expressions with the prognosis of esophageal squamous cell carcinoma (SCC) patients. Ninety-eight cases of esophageal carcinoma patients were enrolled. The expression of KLF4 in the esophageal SCC and normal esophageal mucosa
Hsiao-Fan Chen et al.
Nature communications, 5, 4697-4697 (2014-08-26)
The mechanisms controlling tumour-induced angiogenesis are presently not clear. In principle, angiogenesis can be achieved through the activation of endothelial cells in existing vessels or by transdifferentiation of tumour cells into endothelial cells. However, whether tumour cells can go through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service