Skip to Content
Merck
All Photos(5)

Key Documents

SAB2700363

Sigma-Aldrich

Anti-GPER (C-terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CEPR, CMKRL2, DRY12, GPER

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$2,758.00

R$2,758.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$2,758.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,758.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 1:500-1:3,000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GPER(2852)

Immunogen

Synthetic peptide corresponding to a region within amino acids 307 and 375 of GPR30 according to NP_001035055

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene is a member of the G-protein coupled receptor 1 family and encodes a multi-pass membrane protein that localizes to the endoplasmic reticulum. The protein binds estrogen, resulting in intracellular calcium mobilization and synthesis of phosphatidylinositol 3,4,5-trisphosphate in the nucleus. This protein therefore plays a role in the rapid nongenomic signaling events widely observed following stimulation of cells and tissues with estrogen. Alternate transcriptional splice variants which encode the same protein have been characterized. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayaka Kubota et al.
Biological & pharmaceutical bulletin, 45(8), 1133-1141 (2022-08-01)
Early embryo and sperm transport through the oviductal isthmus depends on the contraction and relaxation of the smooth muscle layers. Dysfunction of the oviduct transport is considered to be one of the causes of infertility. For human infertility, Chinese medicine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service