Skip to Content
Merck
All Photos(2)

Key Documents

SAB2501506

Sigma-Aldrich

Anti-ZCCHC11 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-KIAA0191, Anti-bHLHa22, CCHC domain containing 11, PAP associated domain containing 3, Zinc finger

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, bovine, human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZCCHC11(23318)

General description

Zinc finger CCHC-type containing 11 (ZCCHC11) is a 3′ terminal uridylyl transferase. It possesses multiple domains and has a molecular weight of about 185kDa. A fragment of the protein contains a TRF4 domain, a nucleotidyl transferase domain (NTD), two PAP-associated domains and three zinc finger domains. The ZCCHC11 gene is localized on human chromosome 1p32.3.

Immunogen

Peptide with sequence RERCPHPPRGNVSE, from the C-terminal region of the protein sequence according to NP_001009881.1; NP_056084.1; NP_001009882.1

Biochem/physiol Actions

Zinc finger CCHC-type containing 11 (ZCCHC11) has a role in the RNA-binding protein Lin28 blockade of let-7 microRNA biogenesis. It is recruited by Lin28 to pre-let-7 (let-7 precursors), where it catalyzes the addition of an oligo (U)-tail.The protein is crucial for poly (A) tail length maintenance. ZCCHC11 also has a role in regulating the stability of transcripts for interleukin-6 and other cytokines.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular similarity between myelodysplastic form of chronic
myelomonocytic leukemia and refractory anemia with ring
sideroblasts
Veronique Gelsi-Boyer
Haematologica (2012)
Identification of small molecule inhibitors of Zcchc11 TUTase activity.
Lin S and Gregory RI
RNA Biology, 12(8), 792-800 (2015)
Zcchc11-dependent uridylation of microRNA directs cytokine expression.
Jones MR, et.al
Nature Cell Biology, 11(9), 1157-1163 (2009)
Matthew R Jones et al.
Nature cell biology, 11(9), 1157-1163 (2009-08-25)
Mounting an effective host immune response without incurring inflammatory injury requires the precise regulation of cytokine expression. To achieve this, cytokine mRNAs are post-transcriptionally regulated by diverse RNA-binding proteins and microRNAs (miRNAs) targeting their 3' untranslated regions (UTRs). Zcchc11 (zinc-finger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service