SAB2500629
Anti-Melanocortin 3 Receptor antibody produced in goat
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-MC3, Anti-MC3R
Select a Size
R$2,833.00
Estimated to ship onApril 23, 2025
Select a Size
About This Item
R$2,833.00
Estimated to ship onApril 23, 2025
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
indirect ELISA: suitable
western blot: suitable
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MC3R(4159)
1 of 4
This Item | SAB2501179 | HPA016719 | SAB2500493 |
---|---|---|---|
conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
biological source goat | biological source goat | biological source rabbit | biological source goat |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
technique(s) indirect ELISA: suitable, western blot: suitable | technique(s) indirect ELISA: suitable, western blot: suitable | technique(s) immunohistochemistry: 1:20- 1:50 | technique(s) indirect ELISA: suitable, western blot: suitable |
clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
Immunogen
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service