Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500475

Sigma-Aldrich

Anti-GOT2 (AA 295-306) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Aspartate aminotransferase 2, Anti-FLJ40994, Anti-Glutamic-oxaloacetic transaminase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,833.00

R$2,833.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$2,833.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,833.00


Check Cart for Availability

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOT2(2806)

General description

The GOT2 (glutamic-oxaloacetic transaminase 2) gene is mapped to human chromosome 16q21 and the encoded protein is localized to mitochondria.

Immunogen

Peptide with sequence CKDADEAKRVES from the internal region of the protein sequence according to NP_002071.2.

Biochem/physiol Actions

GOT2 (glutamic-oxaloacetic transaminase 2) participates in malate–aspartate shuttle, which increases the rate of glycolysis via cytosolic NADH (nicotinamide adenine dinucleotide) transfer into mitochondria. This supports rapid growth in tumor cell. GOT2 catalyzes the transamination reaction involving oxaloacetate and L-glutamate to give rise to aspartate, also generating α-ketoglutarate in mitochondria. Aspartate, that is formed, serves as a precursor of N-acetylaspartate, a predominantly abundant free amino acid found in the brain.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association study of got2 genetic polymorphisms and schizophrenia.
Tsai S J, et al.
Psychiatric Genetics, 17(5), 314-314 (2007)
Silvia Sookoian et al.
World journal of gastroenterology, 21(3), 711-725 (2015-01-28)
For several decades, serum levels of alanine (ALT) and aspartate (AST) aminotransferases have been regarded as markers of liver injury, including a wide range of etiologies from viral hepatitis to fatty liver. The increasing worldwide prevalence of metabolic syndrome and
Hui Yang et al.
The EMBO journal, 34(8), 1110-1125 (2015-03-11)
The malate-aspartate shuttle is indispensable for the net transfer of cytosolic NADH into mitochondria to maintain a high rate of glycolysis and to support rapid tumor cell growth. The malate-aspartate shuttle is operated by two pairs of enzymes that localize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service