Skip to Content
Merck
All Photos(7)

Key Documents

SAB2108684

Sigma-Aldrich

Anti-KLF2

affinity isolated antibody

Synonym(s):

Anti-LKLF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,649.00

R$3,649.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,649.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,649.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

pig, human

concentration

0.5-1 mg/mL

technique(s)

ChIP: suitable
immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_016270

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF2(10365)

Immunogen

Synthetic peptide directed towards the middle region of human KLF2

Biochem/physiol Actions

KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.

Sequence

Synthetic peptide located within the following region: PPAFGLFDDAAAAAAALGLAPPAARGLLTPPASPLELLEAKPKRGRRSWP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hairong Chu et al.
Molecular medicine reports, 17(4), 5708-5715 (2018-02-28)
Cardiovascular risk factors can negatively influence late endothelial progenitor cell (EPCs) number and functions, thus EPCs biology is a clinical implications for cardiovascular diseases. The present study aimed to investigate the potential protective effects of resveratrol (RES) on tumor necrosis
Hai-Rong Chu et al.
Molecular medicine reports, 19(3), 1739-1746 (2019-01-11)
The present study aimed to evaluate the effects of Krüppel‑like factor 2 (KLF2) on the differentiation of endothelial progenitor cells (EPCs) to endothelial cells (ECs) induced by shear stress, and to investigate the corresponding mechanisms. Cultured rat late EPCs were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service