Skip to Content
Merck
All Photos(1)

Key Documents

SAB2108408

Sigma-Aldrich

Anti-SLC16A3 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A3(9123)

General description

Solute carrier family 16 member 3 (SLC16A3) encodes monocarboxylate transporter 4 (MCT4). MCT4 is targeted by the chaperone CD147 to the basolateral plasma membrane. MCT4 is highly expressed in astrocytes, skeletal muscle fibres, chondrocytes, placenta. In human chromosome, the gene SLC16A3 is located on 17q25.3.

Immunogen

Synthetic peptide directed towards the N terminal of human SLC16A3

Biochem/physiol Actions

Slc16a3 is a proton-linked monocarboxylate transporter. It catalyses the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate. MCT4 is upregulated in the hypoxic environment in glioblastoma, and is implicated in solid tumours like breast cancer and bladder cancer and leads to metastasis.

Sequence

Synthetic peptide located within the following region: KAVSVFFKELMHEFGIGYSDTAWISSILLAMLYGTGPLCSVCVNRFGCRP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA methylation of the SLC16A3 promoter regulates expression of the human lactate transporter MCT4 in renal cancer with consequences for clinical outcome
Fisel P, et al.
Clinical Cancer Research, 19(18), 5170-5181 (2013)
Identifying differential expression genes and single nucleotide variations using RNA-seq in metastatic melanoma
Liu D, et al.
Genetics and molecular research : GMR, 13, 8153-8162 (2014)
Partial maternal heterodisomy of chromosome 17q25 in a case of severe mental retardation
Rio M, et al.
Human Genetics, 108(6), 511-515 (2001)
Butyric acid increases trans epithelial transport of ferulic acid through upregulation of the monocarboxylate transporters SLC16A1 (MCT1) and SLC16A3 (MCT4)
Ziegler K, et al.
Archives of Biochemistry and Biophysics, 599, 3-12 (2016)
Genetic variations of the MCT4 (SLC16A3) gene in the Chinese and Indian populations of Singapore
Lean CB and Lee EJD
Drug Metabolism and Pharmacokinetics, 27(4), 456-464 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service