Skip to Content
Merck
All Photos(1)

Key Documents

SAB2104191

Sigma-Aldrich

Anti-KIF1A, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ATSV, Anti-C2orf20, Anti-DKFZp686I2094, Anti-FLJ30229, Anti-HUNC-104

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

191 kDa

species reactivity

guinea pig, bovine, mouse, yeast, rat, dog, horse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF1A(547)

Immunogen

Synthetic peptide directed towards the N terminal region of human KIF1A

Biochem/physiol Actions

KIF1A is the motor for anterograde axonal transport of synaptic vesicle precursors.The protein encoded by this gene is a member of the kinesin family. This protein is highly similar to mouse heavy chain kinesin member 1A protein which is an anterograde motor protein that transports membranous organelles along axonal microtubules. It is thought that this protein may play a critical role in the development of axonal neuropathies resulting from impaired axonal transport. There are multiple polyadenylation sites found in this gene. Sequence Note: X90840.1 is a chimeric sequence. Only the ATSV region was propagated into this RefSeq record. [6/26/03, RefSeq staff]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: TTIVNPKQPKETPKSFSFDYSYWSHTSPEDINYASQKQVYRDIGEEMLQH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Neupert et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(40), 13629-13647 (2015-10-09)
Synapses depend on trafficking of key membrane proteins by lateral diffusion from surface populations and by exocytosis from intracellular pools. The cell adhesion molecule neurexin (Nrxn) plays essential roles in synapses, but the dynamics and regulation of its trafficking are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service