Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102436

Sigma-Aldrich

Anti-TJP2 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC26306, Anti-Tight junction protein 2 (zona occludens 2), Anti-X104, Anti-ZO2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 18, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 18, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

118 kDa

species reactivity

rabbit, human, rat, dog, bovine, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TJP2(9414)

General description

The previously assigned protein identifier A2A3H9 has been merged into Q9UDY2. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human TJP2

Biochem/physiol Actions

Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-260 BC027592.1 10-269 261-1975 BC027592.1 270-1984 1976-2543 L27476.1 1855-2422 2544-3142 BC028426.1 2440-3038 3143-4160 L27476.1 3467-4484 4161-4177 BC028426.1 4056-4072

Sequence

Synthetic peptide located within the following region: EPQKAPSRPYQDTRGSYGSDAEEEEYRQQLSEHSKRGYYGQSARYRDTEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service